Isopropanol, 99+%, for spectroscopy
Isopropanol, 99+%, for spectroscopy
Isopropanol, 99+%, for spectroscopy
Isopropanol, 99+%, for spectroscopy
Thermo Scientific Chemicals
Isopropanol, 99+%, for spectroscopy
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1678800101 LGlass Bottle
Catalog number ACR167880010
Price (MYR)
845.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
845.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsopropanol
Melting Point-89.5°C
Absorbance0.01 max. at 300nm, 0.03 max. at 260nm, 0.05 max. at 245nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 1 max. at 205nm
CAS67-63-0
Health Hazard 1Danger
View more
For UV/VIS spectroscopy
RUO – Research Use Only