Chloroform, 99+%, for spectroscopy, stabilized with amylene
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ACR167735000
500 mL
Glass Bottle
Catalog number ACR167735000
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialChloroform
Melting Point-63.0°C
Name NoteStabilized with amylene
Absorbance0.005 max. at 280nm, 0.01 max. at 270nm, 0.075 max. at 260nm, 0.5 max. at 250nm, 1 max. at 245nm
CAS67-66-3