Acetone, 99+%, for spectroscopy
Acetone, 99+%, for spectroscopy
Acetone, 99+%, for spectroscopy
Acetone, 99+%, for spectroscopy
Thermo Scientific Chemicals
Acetone, 99+%, for spectroscopy
CAS: 67-64-1 | C3H6O | 58.08 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1676400252.5 LGlass Bottle
Catalog number ACR167640025
Price (MYR)
951.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
951.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetone
Name Note99+%
Absorbance0.01 max. at 400nm, 0.02 max. at 350nm, 0.1 max. at 340nm, 1 max. at 330nm
CAS67-64-1
Health Hazard 1Danger
View more
For UV/VIS spectroscopy
RUO – Research Use Only