4-tert-Amylphenol, 99%
4-tert-Amylphenol, 99%
4-tert-Amylphenol, 99%
4-tert-Amylphenol, 99%
Thermo Scientific Chemicals
4-tert-Amylphenol, 99%
CAS: 80-46-6 | C11H16O | 164.25 g/mol
Have Questions?
Catalog NumberQuantity
ACR167331000100 g
Catalog number ACR167331000
Price (MYR)
370.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
370.00
EA
Add to cart
Specifications
Chemical Name or Material4-tert-Amylphenol
Name Note99%
CAS80-46-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger