Acetonitrile-d3, for NMR, 99 atom % D
Acetonitrile-d3, for NMR, 99 atom % D
Acetonitrile-d3, for NMR, 99 atom % D
Acetonitrile-d3, for NMR, 99 atom % D
Thermo Scientific Chemicals
Acetonitrile-d3, for NMR, 99 atom % D
Acetonitrile-d3, C2H3N, CAS Number-2206-26-0, 2h3 acetonitrile, acetonitrile-2,2,2-d3, acetonitrile-d3, acetonitrile-d3, >-99.8 atom % d, acetonitrile-d3, 99.8 atom % d, acetonitrile-d isotopic 5g, cd3cn, methyl-d3 cyanide, trideuteroacetonitrile, 10mL, 02, IV, 428, 79.0 deg.C to 81.0 deg.C | CAS: 2206-26-0 | C2H3N | 44.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
25 mL
Packaging:
Glass Bottle
Catalog number ACR166230500
Price (MYR)
2,426.00
BOT
Add to cart
Quantity:
50 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,426.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAcetonitrile-d3
Density0.8440g/mL
Deuterium (D)98.5% min.
Flash Point2°C
PackagingGlass Bottle
View more
For NMR
RUO – Research Use Only