Methyl disulfide, 99%
Methyl disulfide, 99%
Methyl disulfide, 99%
Methyl disulfide, 99%
Thermo Scientific Chemicals
Methyl disulfide, 99%
CAS: 624-92-0 | C2H6S2 | 94.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR165592500250 gGlass bottle
Catalog number ACR165592500
Price (MYR)
450.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
450.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethyl disulfide
CAS624-92-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapour.
Toxic if swallowed.
Toxic if inhaled.
May cause an allergic skin reaction.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
I