Acetyl bromide, 98%
Acetyl bromide, 98%
Acetyl bromide, 98%
Thermo Scientific Chemicals
Acetyl bromide, 98%
CAS: 506-96-7 | C2H3BrO | 122.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ACR159405000
Price (MYR)
2,278.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
2,278.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetyl bromide
Name Note98%
CAS506-96-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger