Acetylcholine Chloride, 99%
Acetylcholine Chloride, 99%
Acetylcholine Chloride, 99%
Acetylcholine Chloride, 99%
Thermo Scientific Chemicals
Acetylcholine Chloride, 99%
CAS: 60-31-1 | C7H16ClNO2 | 181.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number ACR159171000
Price (MYR)
1,360.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,360.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetylcholine chloride
CAS60-31-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0