Cholic acid, 97%
Cholic acid, 97%
Cholic acid, 97%
Thermo Scientific Chemicals
Cholic acid, 97%
CAS: 81-25-4 | C24H40O5 | 408.579 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR159115000
Price (MYR)
3,058.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,058.00
EA
Add to cart
Specifications
Chemical Name or MaterialCholic acid
CAS81-25-4
Health Hazard 2GHS P Statement Causes skin irritation. Causes serious eye irritation.
Health Hazard 3GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Melting Point197°C to 202°C