Pyridinium bromide perbromide, 90+%, technical
Pyridinium bromide perbromide, 90+%, technical
Pyridinium bromide perbromide, 90+%, technical
Pyridinium bromide perbromide, 90+%, technical
Thermo Scientific Chemicals
Pyridinium bromide perbromide, 90+%, technical
CAS: 39416-48-3 | C5H6Br3N | 319.83 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR158361000100 gGlass bottle
Catalog number ACR158361000
Price (MYR)
723.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
723.00
EA
Add to cart
Specifications
Chemical Name or MaterialPyridinium bromide perbromide
Name Note95%
CAS39416-48-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
View more
C-X bond formation reagent
RUO – Research Use Only