2,3-Dimethyl-2-butanol, 98%
2,3-Dimethyl-2-butanol, 98%
2,3-Dimethyl-2-butanol, 98%
2,3-Dimethyl-2-butanol, 98%
Thermo Scientific Chemicals
2,3-Dimethyl-2-butanol, 98%
CAS: 594-60-5 | C6H14O | 102.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR15658025025 mLGlass bottle
Catalog number ACR156580250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 3-Dimethyl-2-butanol
CAS594-60-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0