1,3-Dibromopropane, 98%, stabilized
1,3-Dibromopropane, 98%, stabilized
1,3-Dibromopropane, 98%, stabilized
1,3-Dibromopropane, 98%, stabilized
Thermo Scientific Chemicals
1,3-Dibromopropane, 98%, stabilized
CAS: 109-64-8 | C3H6Br2 | 201.89 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR155041000100 mLGlass bottle
Catalog number ACR155041000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1,3-Dibromopropane
Name Note98%
CAS109-64-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning