3,4-Dichlorophenoxyacetic acid, 96%
3,4-Dichlorophenoxyacetic acid, 96%
3,4-Dichlorophenoxyacetic acid, 96%
3,4-Dichlorophenoxyacetic acid, 96%
Acros Organics
3,4-Dichlorophenoxyacetic acid, 96%
CAS: 588-22-7 | C8H6Cl2O3 | 221.03 g/mol
Have Questions?
Catalog NumberQuantity
ACR1529800505 g
Catalog number ACR152980050
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3, 4-Dichlorophenoxyacetic acid
CAS588-22-7
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0