D(+)-Galactose, 99+%
D(+)-Galactose, 99+%
D(+)-Galactose, 99+%
D(+)-Galactose, 99+%
Thermo Scientific Chemicals
D(+)-Galactose, 99+%
D-Galactose, CAS # 59-23-4, is a right-handed conformation of a monosaccharide that is grouped as an aldohexose, and the molecule may be useful in study of aging, inflammation, and metabolic disorders. | CAS: 59-23-4 | C6H12O6
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ACR150615000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD(+)-Galactose
CAS59-23-4
Melting Point165.0°C to 168.0°C
ColorWhite
Density6.2
View more
General description
  • Galactose is a monosaccharide and aldohexose found in most organisms
  • Galactose is an epimer of glucose and can be converted to glucose through the Leloir pathway
  • It also plays a role as a metabolite in mice and E. coli
  • Galactose is less stable than glucose and rapid conversion often occurs
  • Galactose may also be incorporated into glycoconjugates
Applications
  • Metabolism of galactose may be relevant to the study of aging, inflammation, and metabolic disorders
  • It is intended for a variety of applications from in vitro metabolic assays to HPLC
RUO – Research Use Only