Acetic anhydride, 99+%, pure
Acetic anhydride, 99+%, pure
Acetic anhydride, 99+%, pure
Acetic anhydride, 99+%, pure
Thermo Scientific Chemicals
Acetic anhydride, 99+%, pure
CAS: 108-24-7 | C4H6O3 | 102.089 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Plastic Drum
Catalog number ACR149490250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 L
Packaging:
Plastic Drum
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetic anhydride
Name Note99+%
CAS108-24-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Fatal if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Corrosive to the respiratory tract.
View more
Acylation reagent
RUO – Research Use Only