Propionic acid, 99%, pure
Propionic acid, 99%, pure
Propionic acid, 99%, pure
Propionic acid, 99%, pure
Thermo Scientific Chemicals
Propionic acid, 99%, pure
CAS: 79-09-4 | C3H6O2 | 74.079 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
View all
Glass Bottle
Catalog number ACR149300025
Price (MYR)
766.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
766.00
EA
Add to cart
Specifications
Chemical Name or MaterialPropionic acid
CAS79-09-4
Melting Point-22.0°C
Density0.9900g/mL
Boiling Point141.0°C