Cinnamyl alcohol, 98% trans
Cinnamyl alcohol, 98% trans
Cinnamyl alcohol, 98% trans
Cinnamyl alcohol, 98% trans
Thermo Scientific Chemicals
Cinnamyl alcohol, 98% trans
CAS: 104-54-1 | C9H10O | 134.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1485200505 gGlass bottle
Catalog number ACR148520050
Price (MYR)
387.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
387.00
EA
Add to cart
Specifications
Chemical Name or MaterialCinnamyl alcohol
Name Note98%
CAS104-54-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement