4,4'-Sulfonyldiphenol, 99.7%
4,4'-Sulfonyldiphenol, 99.7%
4,4'-Sulfonyldiphenol, 99.7%
4,4'-Sulfonyldiphenol, 99.7%
Thermo Scientific Chemicals
4,4'-Sulfonyldiphenol, 99.7%
CAS: 80-09-1 | C12H10O4S | 250.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR146915000500 gPlastic bottle
Catalog number ACR146915000
Price (MYR)
923.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
923.00
BOT
Add to cart
Specifications
Chemical Name or Material4, 4'-Sulfonyldiphenol
Name Note99.7%
CAS80-09-1
CAS Min %99.6
Health Hazard 1GHS Signal Word: Warning