Trimethylamine hydrochloride, 98%
Trimethylamine hydrochloride, 98%
Trimethylamine hydrochloride, 98%
Trimethylamine hydrochloride, 98%
Thermo Scientific Chemicals
Trimethylamine hydrochloride, 98%
CAS: 593-81-7 | C3H10ClN | 95.57 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR140071000100 gPlastic bottle
Catalog number ACR140071000
Price (MYR)
389.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
389.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTrimethylamine hydrochloride
Name Note98%
CAS593-81-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: