p-Terphenyl, 99+%, pure
p-Terphenyl, 99+%, pure
p-Terphenyl, 99+%, pure
p-Terphenyl, 99+%, pure
Thermo Scientific Chemicals
p-Terphenyl, 99+%, pure
CAS: 92-94-4 | C18H14 | 230.31 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR137921000100 gPlastic bottle
Catalog number ACR137921000
Price (MYR)
811.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
811.00
EA
Add to cart
Specifications
Chemical Name or Materialp-Terphenyl
Name NotePure
CAS92-94-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement