Sodium hydroxide, 98%, extra pure, pellets
Sodium hydroxide, 98%, extra pure, pellets
Sodium hydroxide, 98%, extra pure, pellets
Sodium hydroxide, 98%, extra pure, pellets
Thermo Scientific Chemicals
Sodium hydroxide, 98%, extra pure, pellets
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1340700101 kgPlastic Bottle
Catalog number ACR134070010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium hydroxide
ColorBeige to Brown
Boiling Point1390.0°C
CAS1310-73-2
Health Hazard 1Danger