Methyl 5-methylsalicylate 97%, ACROS Organics™
Methyl 5-methylsalicylate 97%, ACROS Organics™
Methyl 5-methylsalicylate 97%, ACROS Organics™
Have Questions?
Catalog number ACR133990050
Price (MYR)
-
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl 5-methylsalicylate
Name Note97%
CAS22717-57-3
Health Hazard 2GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more