DL-Serine, 99%
DL-Serine, 99%
DL-Serine, 99%
Thermo Scientific Chemicals
DL-Serine, 99%
CAS: 302-84-1 | C3H7NO3 | 105.093 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR13265025025 gGlass Bottle
Catalog number ACR132650250
Price (MYR)
350.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
350.00
EA
Add to cart
Specifications
Chemical Name or MaterialDL-Serine
Ammonium200ppm max.
CAS302-84-1
ColorWhite
Chlorine (Cl)200ppm max.