Propargyl alcohol, 99%
Propargyl alcohol, 99%
Propargyl alcohol, 99%
Propargyl alcohol, 99%
Thermo Scientific Chemicals
Propargyl alcohol, 99%
CAS: 107-19-7 | C3H4O | 56.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
Glass bottle
Catalog number ACR131455000
Price (MYR)
571.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
571.00
EA
Add to cart
Specifications
Chemical Name or MaterialPropargyl alcohol
Name Note99%
CAS107-19-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Fatal if inhaled.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Suspected o