Piperonylamine, 97%, Thermo Scientific™
Piperonylamine, 97%, Thermo Scientific™
Piperonylamine, 97%, Thermo Scientific™
Piperonylamine, 97%, Thermo Scientific™
Piperonylamine, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR13136025025 mLGlass bottle
Catalog number ACR131360250
Price (MYR)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPiperonylamine
Name Note97%
CAS2620-50-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
View more
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.