Piperonal, 99%
Piperonal, 99%
CAS: 120-57-0 | C8H6O3 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ACR131345000
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPiperonal
CAS120-57-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement May cause an allergic skin reaction.
Health Hazard 3GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.