5-Nitroindole, 99%
5-Nitroindole, 99%
5-Nitroindole, 99%
Thermo Scientific Chemicals
5-Nitroindole, 99%
CAS: 6146-52-7 | C8H6N2O2 | 162.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1287000505 gGlass bottle
Catalog number ACR128700050
Price (MYR)
900.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
900.00
EA
Add to cart
Specifications
Chemical Name or Material5-Nitroindole
Name Note99%
CAS6146-52-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning