Methyltrichlorosilane, 98+%
Methyltrichlorosilane, 98+%
Methyltrichlorosilane, 98+%
Methyltrichlorosilane, 98+%
Thermo Scientific Chemicals
Methyltrichlorosilane, 98+%
CAS: 75-79-6 | CH3Cl3Si | 149.48 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR127935000500 mLGlass bottle
Catalog number ACR127935000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyltrichlorosilane
CAS75-79-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Reacts violently with water.
Corrosive to the res
Health Hazard 3GHS P Statement
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOW