Sulfuric acid, for analysis, ca. 96% solution in water
Sulfuric acid, for analysis, ca. 96% solution in water
Sulfuric acid, for analysis, ca. 96% solution in water
Sulfuric acid, for analysis, ca. 96% solution in water
Thermo Scientific Chemicals
Sulfuric acid, for analysis, ca. 96% solution in water
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1246400101 LGlass Bottle
Catalog number ACR124640010
Price (MYR)
511.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
511.00
EA
Add to cart
Specifications
Chemical Name or MaterialSulfuric acid
CAS7664-93-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
View more
For analysis
RUO – Research Use Only