Perfluoro-compound FC-43(TM), Thermo Scientific Chemicals
Perfluoro-compound FC-43(TM), Thermo Scientific Chemicals
Perfluoro-compound FC-43(TM), Thermo Scientific Chemicals
Perfluoro-compound FC-43(TM), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perfluoro-compound FC-43(TM), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR12377010010 gGlass bottle
Catalog number ACR123770100
Price (MYR)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPerfluoro-compound FC-43*
CAS311-89-7
Melting Point-52.0°C
ColorColorless
Density1.8800g/mL