1,2-Pentanediol, 96%
1,2-Pentanediol, 96%
1,2-Pentanediol, 96%
1,2-Pentanediol, 96%
Thermo Scientific Chemicals
1,2-Pentanediol, 96%
CAS: 5343-92-0 | C5H12O2 | 104.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR123292500250 mLGlass bottle
Catalog number ACR123292500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 2-Pentanediol
Name Note97%
CAS5343-92-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger