2-Ketoglutaric acid, 98%
2-Ketoglutaric acid, 98%
2-Ketoglutaric acid, 98%
2-Ketoglutaric acid, 98%
Thermo Scientific Chemicals
2-Ketoglutaric acid, 98%
CAS: 328-50-7 | C5H6O5 | 146.098 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR12286025025 gPlastic bottle
Catalog number ACR122860250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Ketoglutaric acid
CAS328-50-7
CAS Min %97.5
CAS Max %100.0
Health Hazard 2GHS H Statement: