4-Hydroxybenzyl alcohol, 97%
4-Hydroxybenzyl alcohol, 97%
4-Hydroxybenzyl alcohol, 97%
4-Hydroxybenzyl alcohol, 97%
Thermo Scientific Chemicals
4-Hydroxybenzyl alcohol, 97%
CAS: 623-05-2 | C7H8O2 | 124.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR121031000100 gGlass bottle
Catalog number ACR121031000
Price (MYR)
1,448.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,448.00
EA
Add to cart
Specifications
Chemical Name or Material4-Hydroxybenzyl alcohol
CAS623-05-2
Health Hazard 1GHS Signal Word: Warning
Melting Point110.0°C to 112.0°C
ColorBeige to Pink