Dioctyl phthalate, 99%, ACROS Organics™
Dioctyl phthalate, 99%, ACROS Organics™
Dioctyl phthalate, 99%, ACROS Organics™
Have Questions?
Catalog number ACR117091000
Price (MYR)
-
Request bulk or custom format
Specifications
Chemical Name or MaterialDioctyl phthalate
CAS117-81-7
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
May damage fertility. May damage the unborn child.
Health Hazard 3GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more