2,5-Dimethylthiophene, 98.5%
2,5-Dimethylthiophene, 98.5%
2,5-Dimethylthiophene, 98.5%
2,5-Dimethylthiophene, 98.5%
Thermo Scientific Chemicals
2,5-Dimethylthiophene, 98.5%
CAS: 638-02-8 | C6H8S | 112.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR11688025025 mLGlass bottle
Catalog number ACR116880250
Price (MYR)
1,132.00
EA
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,132.00
EA
Add to cart
Specifications
Chemical Name or Material2, 5-Dimethylthiophene
Name Note98.5%
CAS638-02-8
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning