Wide-Mouth 1000 mL Wash Bottles
Wide-Mouth 1000 mL Wash Bottles
Actual product may vary
Thermo Scientific Chemicals
Wide-Mouth 1000 mL Wash Bottles
These wide-mouth wash bottles are made of natural polyethylene and come with a cap.
Have Questions?
Catalog NumberCapacity (Metric)
ACR1162010001000 mL
Catalog number ACR116201000
Price (MYR)
826.00
BOT
Add to cart
Capacity (Metric):
1000 mL
Price (MYR)
826.00
BOT
Add to cart
These wide-mouth wash bottles are made of natural polyethylene and come with a cap.
  • White wash bottle
  • Made of low-density polyethylene (LDPE)
  • Color-coded polypropylene closures for easy identification
  • Wide-mouths for easy filling
Specifications
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorWhite
MaterialLDPE
Quantity3
SterilityNon-sterile
Closure IncludedYes
AutoclavableNot Autoclavable
DisposableReusable
GraduatedNo
MouthWide
Height (Metric)210 mm
Product TypeLDPE Wash Bottles
Closure Size53 mm
Diameter (Metric)95 mm
Closure TypeSingle Screw Cap
Unit SizeBOT