4,6-Dihydroxy-2-mercaptopyrimidine, 98%
4,6-Dihydroxy-2-mercaptopyrimidine, 98%
4,6-Dihydroxy-2-mercaptopyrimidine, 98%
4,6-Dihydroxy-2-mercaptopyrimidine, 98%
Thermo Scientific Chemicals
4,6-Dihydroxy-2-mercaptopyrimidine, 98%
CAS: 504-17-6 | C4H4N2O2S | 144.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number ACR114965000
Price (MYR)
2,463.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,463.00
EA
Add to cart
Specifications
Chemical Name or Material4, 6-Dihydroxy-2-mercaptopyrimidine
Name Note98%
CAS504-17-6
CAS Min %97.5
Health Hazard 1Warning