2',4'-Dihydroxyacetophenone, 98%
2',4'-Dihydroxyacetophenone, 98%
2',4'-Dihydroxyacetophenone, 98%
2',4'-Dihydroxyacetophenone, 98%
Thermo Scientific Chemicals
2',4'-Dihydroxyacetophenone, 98%
CAS: 89-84-9 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
View all
Plastic bottle
Catalog number ACR114801000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2', 4'-Dihydroxyacetophenone
Name Note98%
CAS89-84-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning