cis-1,2-Dichloroethylene, 97%, stabilized, Thermo Scientific Chemicals
cis-1,2-Dichloroethylene, 97%, stabilized, Thermo Scientific Chemicals
cis-1,2-Dichloroethylene, 97%, stabilized, Thermo Scientific Chemicals
cis-1,2-Dichloroethylene, 97%, stabilized, Thermo Scientific Chemicals
cis-1,2-Dichloroethylene, 97%, stabilized, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
ACR11338050050 mLGlass bottle
Catalog number ACR113380500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialcis-1, 2-Dichloroethylene
Name Note97%
CAS156-59-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.