Check Order Status
MicroTee
Actual product may vary
Thermo Scientific Chemicals
MicroTee
Connects 3 capillary tubes, using MicroFingertight II fittings. Includes 3 MicroFerrules for 0.025 in. O.D. tubing, 3 female nuts.
Have Questions?
Change view
Catalog Number
For Use With
ACR111010250
360 μm O.D. Tubing
FSA111010250
360 μm O.D. Tubing
2 Options
Catalog number
ACR111010250
Price (MYR)
1,063.00
EA
Add to cart
For Use With:
360 μm O.D. Tubing
Price (MYR)
1,063.00
EA
Add to cart
Specifications
Packaging Quantity
1/Pk.
Includes
3, microferrules for 360 μm O.D. tubing and 3, female nuts (5/16 in. - 24 internal threads)
For Use With
360 μm O.D. Tubing