sec-Butylbenzene, 99+%
sec-Butylbenzene, 99+%
sec-Butylbenzene, 99+%
sec-Butylbenzene, 99+%
Thermo Scientific Chemicals
sec-Butylbenzene, 99+%
CAS: 135-98-8 | C10H14 | 134.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1078600505 mLGlass bottle
Catalog number ACR107860050
Price (MYR)
343.00
EA
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
343.00
EA
Add to cart
Specifications
Chemical Name or Materialsec-Butylbenzene
Name Note99+%
CAS135-98-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.