2-Bromobenzonitrile, 99%
2-Bromobenzonitrile, 99%
2-Bromobenzonitrile, 99%
2-Bromobenzonitrile, 99%
Thermo Scientific Chemicals
2-Bromobenzonitrile, 99%
CAS: 2042-37-7 | C7H4BrN | 182.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR10669010010 gGlass bottle
Catalog number ACR106690100
Price (MYR)
834.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
834.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Bromobenzonitrile
Name Note99%
CAS2042-37-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning