Benzamidine Hydrochloride Hydrate, 98%
Benzamidine Hydrochloride Hydrate, 98%
Benzamidine Hydrochloride Hydrate, 98%
Benzamidine Hydrochloride Hydrate, 98%
Thermo Scientific Chemicals
Benzamidine Hydrochloride Hydrate, 98%
CAS: 206752-36-5 | C7H9ClN2 | 156.61 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR1052400505 gGlass Bottle
Catalog number ACR105240050
Price (MYR)
388.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
388.00
EA
Add to cart
Specifications
Chemical Name or MaterialBenzamidine hydrochloride hydrate
CAS206752-36-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0