Anthranilamide, 99+%
Anthranilamide, 99+%
Anthranilamide, 99+%
Anthranilamide, 99+%
Thermo Scientific Chemicals
Anthranilamide, 99+%
CAS: 88-68-6 | C7H8N2O | 136.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR104901000100 gPlastic bottle
Catalog number ACR104901000
Price (MYR)
434.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
434.00
EA
Add to cart
Specifications
Chemical Name or MaterialAnthranilamide
CAS88-68-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.