Allyl isothiocyanate, 94%, stabilized with 0.01% alpha-tocopherol
Allyl isothiocyanate, 94%, stabilized with 0.01% alpha-tocopherol
Allyl isothiocyanate, 94%, stabilized with 0.01% alpha-tocopherol
Allyl isothiocyanate, 94%, stabilized with 0.01% alpha-tocopherol
Thermo Scientific Chemicals
Allyl isothiocyanate, 94%, stabilized with 0.01% alpha-tocopherol
CAS: 57-06-7 | C4H5NS | 99.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR102955000
Price (MYR)
1,259.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,259.00
EA
Add to cart
Specifications
Chemical Name or MaterialAllyl isothiocyanate
Name Note94%
CAS57-06-7
CAS Min %92.0
Health Hazard 1Danger