Abgene™ 96 Well 2.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Abgene™ 96 Well 2.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Actual product may vary
Abgene™ 96 Well 2.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Abgene™ 96 Well 2.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Abgene™ 96 Well 2.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Abgene™ 96 Well 2.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Abgene™ 96 Well 2.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
2.2 mL deepwell storage plates for the culture or storage of large samples.
Have Questions?
Catalog NumberColorWell ShapePackagingFit for Purpose
AB-0932NaturalSquare, Conical Bottom (V-well)Individually WrappedPCR Ready
Catalog number AB-0932
Price (MYR)
1,737.00
CS
Add to cart
Color:
Natural
Well Shape:
Square, Conical Bottom (V-well)
Packaging:
Individually Wrapped
Fit for Purpose:
PCR Ready
Price (MYR)
1,737.00
CS
Add to cart
Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
Highlights
  • Designed for optimal sample recovery
  • Each square well is specifically designed with an independent sealing rim to prevent cross-contamination
  • U-bottom (AB-0661) to aid resuspension or conical bottom (AB-0932) for ease of sample retrieval
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceCertified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
FormatStandard
Description96-well Polypropylene Sample Processing & Storage Plates
Array8 x 12
AutoclavableAutoclavable
PackagingIndividually Wrapped
Binding TypeLow binding PP
Product LineAbgene
ColorNatural
Product Type96-well plate
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
MaterialPolypropylene
Volume (Metric) Well2200 μL
No. of Wells96
Volume (Metric) Working1800 μL
SterilityCleanroom
Surface TreatmentNon-treated
Well ShapeSquare, Conical Bottom (V-well)
Dnase FreeYes
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
Max. RCF5000 x g
No. per Case50
No. per Pack1
Quantity50/Cs.
Regulatory StatusFor Research Use Only. Not for use in diagnostic procedures.
RNase FreeYes
Fit for PurposePCR Ready
Plate SizeANSI/ SLAS Standard
Unit SizeCS