Reclosable Poly Bags
Reclosable Poly Bags
Actual product may vary
Reclosable Poly Bags
Clear, resealable polyethylene bags
Have Questions?
Catalog NumberDimensions (L x W)Thickness (English)
A3-113 x 10 in. (33.02 x 25.4cm)2 mil
Catalog number A3-1
Price (MYR)
9,727.00
EA
Add to cart
Dimensions (L x W):
13 x 10 in. (33.02 x 25.4cm)
Thickness (English):
2 mil
Price (MYR)
9,727.00
EA
Add to cart
  • Thickness: 2 or 4mil
  • Seal securely with zipper lock
Specifications
ColorClear
For Use With (Application)Provides protection for contents and clear visibility
MaterialPolyethylene
Thickness (Metric)0.279 mm
Thickness (English)2 mil
Dimensions (L x W)13 x 10 in. (33.02 x 25.4cm)
Length (English)13 in.
Length (Metric)33.02 cm
Width (English)10 in.
Width (Metric)25.4 cm
Product TypeReclosable Poly Bag
Unit SizeEA
Frequently asked questions (FAQs)
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.