1 mL Tuberculin Luer Slip Syringe
1 mL Tuberculin Luer Slip Syringe
Actual product may vary
1 mL Tuberculin Luer Slip Syringe
Inert syringes for use in gas or liquid chromatography, in vitro fertilization, embryo-transfer and various laboratory procedures
Have Questions?
Catalog NumberdimNeedleGauge
A1
Catalog number A1
Price (MYR)
4,758.00
EA
Add to cart
Price (MYR)
4,758.00
EA
Add to cart
  • Needle tips not included, purchased separately
  • Graduated syringes manufactured from high-quality laboratory-grade polypropylene and polyethylene
  • Chemically inert and noncytotoxic
  • Ideal for chromatography, in vitro fertilization and embryo transfer
  • Positive safety stop prevents accidental spills
  • FDA registered
Over the coming months, you may notice a difference in the box you receive as a branding transition occurs with the Norm-Ject line of syringes. These syringes continue to be manufactured in the same location and the same high standards. Please contact us for more details.
Specifications
Certifications/ComplianceFDA, CE, ISO 9001, DEHP free
MaterialPlastic
For Use With (Application)For nuclear medicine, amniocentesis, IVF, embryo-transfer, chromatography and lab procedures
Latex FreeYes
Product TypeB Braun syringe
Graduations0.05 cc
Quantity100 Pk., 18 Pk./Cs.
PackagingIndividually wrapped
Volume (Metric)1 mL
Product LineB Braun
Needle Point StyleCone
Connection TypeLeur Slip