KingFisher™ Plastics for 96 deep-well format
KingFisher™ Plastics for 96 deep-well format
Actual product may vary
KingFisher™ Plastics for 96 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
97003530Combi PackKingFisher Duo Prime only
9504045096 Deep Well plate, v-bottomKF Duo Prime, Flex and Presto
Catalog number 97003530
Price (MYR)
-
Product Type:
Combi Pack
For Use With (Equipment):
KingFisher Duo Prime only
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Product TypeCombi Pack
Format96 Deep-well Plate
DescriptionKingFisher combi pack for microtiter 96 deep-well plate (includes 8 plates, 8 tip combs, and 8 elution strips and caps) (for Duo Prime only).
For Use With (Equipment)KingFisher Duo Prime only
Unit SizeEA
Frequently asked questions (FAQs)
How many plates come with the KingFisher combi pack for microtiter 96 deep-well plate (Cat. No. 97003530)?
This kit is for the KingFisher Duo Prime only. It contains the following for processing 96 samples:
• 8 plates
• 8 tip combs
• 8 elution strips
• 8 elution strip caps
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
I received the KingFisher Flex/KingFisher Duo Prime tip combs and the retaining clips are bent or broken off. When I try to unbend the retaining clips, they break. Can I still use the tip combs if the retaining clips are bent or broken off?
Do not use the tip combs if the clips are bent or missing. Not having clips can cause the tip comb to drop from the holder during the run, which would cause the run to abort. If you received tip combs with bent or broken retaining clips, you can contact Technical Support at techsupport@thermofisher.com to request a free-of-charge replacement.
Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.
What kind of consumables do I need for the KingFisher Duo Purification System?
The KingFisher Duo Purification System consumables are listed on the following website and they vary depending on whether you are using the 6- or 12-pin magnet: https://www.thermofisher.com/order/catalog/product/97003500
What is the difference between the KingFisher Duo and the KingFisher Duo Prime Purification Systems?
The KingFisher Duo Prime Purification System has the following additional features compared to the KingFisher Duo Purification System:
• Improved safety shield design
• UV lamp
• Handheld barcode reader option
• Additional language options
• Optimized elution strip block for better alignment of the plastic strip
The KingFisher Duo can run any script written for the KingFisher Duo Prime and does not require additional modifications.
Are KingFisher Plastics for 96-deep well format autoclavable?
No, KingFisher Plastics for 96-deep well format are not autoclavable. They won't melt, but they will get deformed.