KingFisher™ Plastics for 96 standard and PCR formats
KingFisher™ Plastics for 96 standard and PCR formats
Actual product may vary
KingFisher™ Plastics for 96 standard and PCR formats
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Have Questions?
Catalog NumberFor Use With (Equipment)Quantity
97002540KingFisher Flex and Presto48/Cs.
Catalog number 97002540
Price (MYR)
3,014.00
PK
Add to cart
For Use With (Equipment):
KingFisher Flex and Presto
Quantity:
48/Cs.
Price (MYR)
3,014.00
PK
Add to cart
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Format96 Deep-well Plate
Capacity96 Samples
DescriptionKingFisher 96 microplate (200μL) (for Flex and Presto)
*Note: Only works with KingFisher™ Flex 96 Heating Block (Cat. No. 24075420). Not to be used with KingFisher™ Flex 96 Deep-Well Heating Block (Cat. No. 24075430).
For Use With (Equipment)KingFisher Flex and Presto
Quantity48/Cs.
Unit SizePK
Frequently asked questions (FAQs)
Are the MagMAX Express and KingFisher plates the same?
Yes, the MagMAX Express and KingFisher plates are equivalent. We recommend the KingFisher plates as an alternative to the discontinued MagMAX Express plates.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.
Can I use the same tips and plates for the KingFisher Flex instrument with the 96-well magnetic head and the MagMAX Express-96 instrument magnetic head?
Yes, the KingFisher Flex instrument 96-well magnetic head and the MagMAX Express-96 instrument magnetic head can use the same tip combs and KingFisher plates even though the magnetic heads are different.
Do you offer an alternative for the discontinued Pharma MagMAX Express-96 Standard Plates (Cat. No. A31541)?
Yes, we recommend using KingFisher 96 microplate (for KingFisher Flex, Apex, and Presto) (Cat. No. 97002540).
Note: Only works with KingFisher Flex 96 Heating Block (Cat. No. 24075420). Not to be used with KingFisher Flex 96 Deep-Well Heating Block (Cat. No. 24075430).
Do I have to buy any additional consumables along with the MagMAX kits if I want to use them on the KingFisher Flex instrument?
Yes, depending on the kind of protocol and head that is used, the following consumables will have to be purchased:
- KingFisher 96 deep-well plate (Cat. No. 95040450)
- KingFisher 96 deep-well plate, sterile (Cat. No. 95040460)
- KingFisher 96 tip comb for deep-well (Cat. No. 97002534)
- KingFisher 96 deep-well tip comb and plate, sterile (Cat. No. 97002820)
- KingFisher 96 microplate (200 μL) (Cat. No. 97002540)
- KingFisher 96 tip comb (Cat. No. 97002524)
- MicroAmp Clear Adhesive Film (optional) (Cat. No. 4306311
Can the KingFisher Flex 96 KF and 96 Deep-Well heating blocks be used for a KingFisher Flex Magnetic Particle Processor with a 96 Deep-Well Head?
Yes, both the KingFisher Flex 96 KF and 96 Deep-Well heating blocks can be used for the 96 Deep-Well Head, but they are only compatible with certain MagMAX/KingFisher plates.
- The KingFisher Flex 96 KF Heating Block is compatible with both standard plates and Deep-Well plates (Cat. Nos. 97002540, 4388475, 95040460, 4388476).
- The KingFisher Flex Deep-Well Heating Block is only compatible with Deep-Well plates (Cat. Nos. 95040460, 4388476).